Learn More
Enzo Life Sciences BIOARRAY™ methylated DNA IP Kit (10tests)
SDP
Catalog No. 502012570
Encompass_Preferred
The BIOARRAY™ methylated DNA IP kit features IP technology for the enrichment of 5-methylcytosine-containing DNA from any pool of fragmented genomic DNA for use in genome-wide methylation analysis. The kit includes a highly specific anti-5-methylcytosine monoclonal antibody for the “capture” and separation of methylated DNA from non-methylated DNA in only a few hours.
Catalog No. 50-201-2570 Supplier Enzo Life Sciences Supplier No. ENZ450120010
The BIOARRAY™ methylated DNA IP kit features IP technology for the enrichment of 5-methylcytosine-containing DNA from any pool of fragmented genomic DNA for use in genome-wide methylation analysis. The kit includes a highly specific anti-5-methylcytosine monoclonal antibody for the “capture” and separation of methylated DNA from non-methylated DNA in only a few hours.