Learn More
Biochain Institute Inc Methylated & Low-Methylated DNA Matched Pair - Human Peripheral Blood Leukocyte, 2 x 5 ug/pk
SDP
Catalog No. 501871521
Encompass_Preferred
The methylated and low-methylated matched pair DNA can be utilized as positive and negative controls,respectively, in research focusing on gene methylation studies. The low-methylated genomic DNAs are selected from human young donor blood peripheral leukocyte, liver tissue, or lymphocyte cell line. The DNAs derived from these sources naturally have a low level of methylation and display batch-to-batch consistency. Unlike DNAs from the DNA methyltransferase genetic knockout cell lines, these DNA sources are ideal for more natural control DNAs to be used in gene methylation studies.
Catalog No. 50-187-1521 Supplier Biochain Institute Inc Supplier No. D6234148PP
The methylated and low-methylated matched pair DNA can be utilized as positive and negative controls,respectively, in research focusing on gene methylation studies. The low-methylated genomic DNAs are selected from human young donor blood peripheral leukocyte, liver tissue, or lymphocyte cell line. The DNAs derived from these sources naturally have a low level of methylation and display batch-to-batch consistency. Unlike DNAs from the DNA methyltransferase genetic knockout cell lines, these DNA sources are ideal for more natural control DNAs to be used in gene methylation studies.