Learn More
Active Motif Methylated DNA Standard Kit
SDP
Catalog No. NC1852696
Encompass
Active Motif's Methylated DNA Standard Kit includes three recombinant DNA standards derived from the APC gene promoter: unmethylated DNA, 5-methylcytosine methylated DNA and 5-hydroxymethylcytosine methylated DNA. This kit (which also includes PCR primers specific to the APC promoter) can be used as part of experiments utilizing our antibodies specific to the different types of DNA methylation.
Catalog No. NC1852696 Supplier Active Motif Supplier No. 55008
Active Motif's Methylated DNA Standard Kit includes three recombinant DNA standards derived from the APC gene promoter: unmethylated DNA, 5-methylcytosine methylated DNA and 5-hydroxymethylcytosine methylated DNA. This kit (which also includes PCR primers specific to the APC promoter) can be used as part of experiments utilizing our antibodies specific to the different types of DNA methylation.