Learn More
Cayman Chemical Methylated LysIn PolycLNal
SDP
Catalog No. 502955054
Encompass_Preferred
Immunogen: Methylated KLH • Host: Rabbit • Species reactivity: (+) Species independent • Applications: ELISA, ICC, IF, IP, WB • Lysine can be methylated once, twice, or three times by lysine methyltransferases. The transfer of methyl groups from SAM to histones is catalyzed by histone methyltransferases.
Catalog No. 50-295-5054 Supplier Cayman Chemical Supplier No. 13729100 UG
Immunogen: Methylated KLH • Host: Rabbit • Species reactivity: (+) Species independent • Applications: ELISA, ICC, IF, IP, WB • Lysine can be methylated once, twice, or three times by lysine methyltransferases. The transfer of methyl groups from SAM to histones is catalyzed by histone methyltransferases.