Learn More
Zymo Research Corporation Methylated/Non-methylated Control DNA & Primer Set, 1 set
SDP
Catalog No. 502037134
Encompass_Preferred
The Control DNA and Control Primers provided with it can be used for monitoring the different steps of the methylated-DNA IP procedure. The pUC19m DNA contains a mutation to create a novel Nco I restriction enzyme site. Since the methylated DNA (pUC19m) contains a Nco I restriction site (the non-methylated (pUC19) DNA does not), Nco I digestion can be used to differentiate between methylated and non-methylated DNAs.
Catalog No. 50-203-7134 Supplier Zymo Research Corporation Supplier No. D51012
The Control DNA and Control Primers provided with it can be used for monitoring the different steps of the methylated-DNA IP procedure. The pUC19m DNA contains a mutation to create a novel Nco I restriction enzyme site. Since the methylated DNA (pUC19m) contains a Nco I restriction site (the non-methylated (pUC19) DNA does not), Nco I digestion can be used to differentiate between methylated and non-methylated DNAs.