Learn More
Cayman Chemical Methylatropine (nitrate), Purity: ≥95%, CAS: 52-88-0, 100 mg
SDP
Catalog No. 502892709
Encompass_Preferred
A muscarinic acetylcholine antagonist (IC50 = 50 = 5.5 µg/kg); reduces salivation, induces mydriasis, and increases heart rate in dogs
Catalog No. 50-289-2709 Supplier Cayman Chemical Supplier No. 9002272100 MG
A muscarinic acetylcholine antagonist (IC50 = 50 = 5.5 µg/kg); reduces salivation, induces mydriasis, and increases heart rate in dogs