Learn More
Cayman Chemical Methylatropine (nitrate), Purity: ≥95%, CAS: 52-88-0, 50 mg
SDP
Catalog No. 502892534
Encompass_Preferred
A muscarinic acetylcholine antagonist (IC50 = 50 = 5.5 µg/kg); reduces salivation, induces mydriasis, and increases heart rate in dogs
Catalog No. 50-289-2534 Supplier Cayman Chemical Supplier No. 900227250 MG
A muscarinic acetylcholine antagonist (IC50 = 50 = 5.5 µg/kg); reduces salivation, induces mydriasis, and increases heart rate in dogs