Learn More
FUJIFILM Biosciences Methylazxymethanl Acetate MAM
Catalog No. 5000450211
Encompass_Preferred
Methylazoxymethanol Acetate MAM is used in preparation of model animals of schizophrenia as it reduces neurogenesis when administered in rats etc In patients with psychiatric disorders such as schizophrenia attenuation of sensory filter function that shuts unnecessary noises in the surroundings out of consciousness is observed This sensory filter function may be evaluated by a physiological test known as prepulse inhibition PPI weakening reaction to frightening sounds The decrease in this PPI is correlated to decrease in neurogenesis For research use only RUO
Catalog No. 50-004-50211 Supplier FUJIFILM Biosciences Supplier No. 13616303
Methylazoxymethanol Acetate MAM is used in preparation of model animals of schizophrenia as it reduces neurogenesis when administered in rats etc In patients with psychiatric disorders such as schizophrenia attenuation of sensory filter function that shuts unnecessary noises in the surroundings out of consciousness is observed This sensory filter function may be evaluated by a physiological test known as prepulse inhibition PPI weakening reaction to frightening sounds The decrease in this PPI is correlated to decrease in neurogenesis For research use only RUO