Learn More
Medchemexpress LLC HY-B2070 50mg Medchemexpress, Methylbenactyzium Bromide CAS:3166-62-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB207050MG
Encompass_Preferred
Medchemexpress, HY-B2070 50mg Methylbenactyzium Bromide CAS:3166-62-9 Methylbenactyzium Bromide is a muscarinic acetylcholine receptor ( mAChR ) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3057