Learn More
Medchemexpress LLC Methylbenactyzium bromide | 3166-62-9 | 99.53% | 200 MG
SDP
Catalog No. 5000410587
Encompass_Preferred
Methylbenactyzium Bromide is a muscarinic acetylcholine receptor (mAChR) inhibitor. It has been used as a spasmolytic for the treatment of gastrointestinal ulcer.
  • Muscarinic acetylcholine receptor (mAChR) inhibitor.
  • Used as a spasmolytic for the treatment of gastrointestinal ulcer.
Catalog No. 50-004-10587 Supplier Medchemexpress LLC Supplier No. HYB2070200MG
Methylbenactyzium Bromide is a muscarinic acetylcholine receptor (mAChR) inhibitor. It has been used as a spasmolytic for the treatment of gastrointestinal ulcer.
  • Muscarinic acetylcholine receptor (mAChR) inhibitor.
  • Used as a spasmolytic for the treatment of gastrointestinal ulcer.