Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Methylbenzyl Cyanide 96%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
50g
1823-91-2
C9H9N
131.178
MFCD00001871
NVAOLENBKNECGF-UHFFFAOYSA-N
2-phenylpropionitrile,hydratroponitrile,alpha-methylphenylacetonitrile,alpha-methylbenzyl cyanide,2-phenylpropiononitrile,benzeneacetonitrile, .alpha.-methyl,methylbenzyl cyanide,.alpha.-phenylpropionitrile,.alpha.-phenylethyl cyanide,.alpha.-methylbenzyl cyanide
15761
2-phenylpropanenitrile
CC(C#N)C1=CC=CC=C1
1823-91-2
C9H9N
131.178
MFCD00001871
NVAOLENBKNECGF-UHFFFAOYSA-N
2-phenylpropionitrile,hydratroponitrile,alpha-methylphenylacetonitrile,alpha-methylbenzyl cyanide,2-phenylpropiononitrile,benzeneacetonitrile, .alpha.-methyl,methylbenzyl cyanide,.alpha.-phenylpropionitrile,.alpha.-phenylethyl cyanide,.alpha.-methylbenzyl cyanide
15761
2-phenylpropanenitrile
CC(C#N)C1=CC=CC=C1
Infrared Spectrum Authentic
Color Yellow
Density 0.98g/mL
Refractive Index 1.5096 to 1.5116 (20°C, 589nm)
Quantity 25g
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Linear Formula C6H5CH(CH3)CN
Formula Weight 131.18
Physical Form Liquid
Percent Purity ≥95%
Chemical Name or Material α-Methylbenzyl cyanide, 96%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 217-354-9

TSCA : TSCA