Learn More
Medchemexpress LLC Methylboronic acid | 13061-96-6 | ≥98.0% | 59.86 | 100 G
SDP
Catalog No. 5000403654
Encompass_Preferred
Methylboronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is strictly for research use only and not sold to patients.
  • Suitable for various life science research applications
  • Appears as a white to off-white solid
  • Soluble in DMSO for in vitro studies
  • Requires storage at -20°C under nitrogen
Catalog No. 50-004-03654 Supplier Medchemexpress LLC Supplier No. HY34449100G
Methylboronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is strictly for research use only and not sold to patients.
  • Suitable for various life science research applications
  • Appears as a white to off-white solid
  • Soluble in DMSO for in vitro studies
  • Requires storage at -20°C under nitrogen