Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methylboronic acid | 13061-96-6 | ≥98.0% | 59.86 | 25 G
SDP

Catalog No. 5000403079
Encompass_Preferred

Methylboronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only and not sold to patients.

  • Purity: ≥98.0%
  • CAS number: 13061-96-6
  • Molecular weight: 59.86
  • Molecular formula: CH₅BO₂
  • Appearance: White to off-white solid
  • Storage: -20°C, stored under nitrogen
  • Solubility in DMSO: 100 mg/mL

Catalog No. 50-004-03079 Supplier Medchemexpress LLC Supplier No. 25GHY34449
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methylboronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only and not sold to patients.

  • Purity: ≥98.0%
  • CAS number: 13061-96-6
  • Molecular weight: 59.86
  • Molecular formula: CH₅BO₂
  • Appearance: White to off-white solid
  • Storage: -20°C, stored under nitrogen
  • Solubility in DMSO: 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.