Learn More
Medchemexpress LLC Methylboronic acid-d3 | 1332481-37-4 | 99.8% | 62.88 | 5 MG
SDP
Catalog No. 5000401503
Encompass_Preferred
Methylboronic acid-d3 is a stable isotope-labeled compound, primarily used in research applications. This deuterated form of methylboronic acid is valuable in various scientific studies, including those requiring the use of stable isotopes for metabolic tracing, quantitative analysis, or reaction mechanism elucidation. Its high purity ensures reliable and consistent results in sensitive analytical techniques.
  • Labeled with deuterium for research applications.
  • Suitable for use as a tracer in biological and chemical studies.
  • Utilized as an internal standard in analytical techniques.
  • Offers high purity for accurate experimental outcomes.
  • Ideal for investigations into reaction mechanisms.
Catalog No. 50-004-01503 Supplier Medchemexpress LLC Supplier No. HY34449S5MG
Methylboronic acid-d3 is a stable isotope-labeled compound, primarily used in research applications. This deuterated form of methylboronic acid is valuable in various scientific studies, including those requiring the use of stable isotopes for metabolic tracing, quantitative analysis, or reaction mechanism elucidation. Its high purity ensures reliable and consistent results in sensitive analytical techniques.
  • Labeled with deuterium for research applications.
  • Suitable for use as a tracer in biological and chemical studies.
  • Utilized as an internal standard in analytical techniques.
  • Offers high purity for accurate experimental outcomes.
  • Ideal for investigations into reaction mechanisms.