Learn More
Abcam Methylcarbamyl-PAF, non-hydrolyzable platelet-activating factor (PAF) analog, 5MG
Catalog No. 502706481
Encompass_Preferred
MW 538.7 Da, Purity >=98%. Non-hydrolyzable platelet-activating factor (PAF) analog. Causes fetal and placental growth restriction. Alters mitochondrial membrane. Induced vascular VEGF expression in cord endothelial cells. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6481 Supplier Abcam Supplier No. AB1416815MG
MW 538.7 Da, Purity >=98%. Non-hydrolyzable platelet-activating factor (PAF) analog. Causes fetal and placental growth restriction. Alters mitochondrial membrane. Induced vascular VEGF expression in cord endothelial cells. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement