Learn More
Cayman Chemical Methylcardol trIen 10mg
SDP
Catalog No. 502987857
Encompass_Preferred
A phenol from cashew nut shell liquid; inhibits α-glucosidase (IC50 = 39.6 µM); kills 100% of adult S. mansoni worms after 24 hours when used at concentrations of 100 and 200 µM; has been used as a starting material for the synthesis of mono- and bis-benzoxazines
Catalog No. 50-298-7857 Supplier Cayman Chemical Supplier No. 2340410 MG
A phenol from cashew nut shell liquid; inhibits α-glucosidase (IC50 = 39.6 µM); kills 100% of adult S. mansoni worms after 24 hours when used at concentrations of 100 and 200 µM; has been used as a starting material for the synthesis of mono- and bis-benzoxazines