Learn More
Medchemexpress LLC MethylCBI-azaindole-benzamide-MOM-Boc-ethylenediamine-D, 25mg
SDP
Catalog No. NC3781111
Encompass_Preferred
MethylCBI-azaindole-benzamide-MOM-Boc-ethylenediamine-D is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs).
Catalog No. NC3781111 Supplier Medchemexpress LLC Supplier No. HY14548825MG
Add to Cart
Add to Cart
Request a Quote
MethylCBI-azaindole-benzamide-MOM-Boc-ethylenediamine-D is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs).