Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methylcbi-azaindole-benzamide-mom-boc-ethylenediamine-d | 2222445-87-4 | 99.7% | 859.36 | C44H51ClN6O10 | 1 MG
SDP

Catalog No. 5000202642
Encompass_Preferred

MethylCBI-azaindole-benzamide-MOM-Boc-ethylenediamine-D is a cleavable ADC linker used in the synthesis of antibody-drug conjugates and supplied as a high-purity research reagent for conjugation and synthetic workflows.

  • Cleavable linker for intracellular release
  • High purity (99.7%)
  • Molecular weight 859.36 g/mol
  • Available in small research quantities (1 mg and larger)
  • Storage: powder at -20°C; in solvent at -80°C (6 months) or -20°C (1 month)

Catalog No. 50-002-02642 Supplier Medchemexpress LLC Supplier No. HY1454881MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MethylCBI-azaindole-benzamide-MOM-Boc-ethylenediamine-D is a cleavable ADC linker used in the synthesis of antibody-drug conjugates and supplied as a high-purity research reagent for conjugation and synthetic workflows.

  • Cleavable linker for intracellular release
  • High purity (99.7%)
  • Molecular weight 859.36 g/mol
  • Available in small research quantities (1 mg and larger)
  • Storage: powder at -20°C; in solvent at -80°C (6 months) or -20°C (1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.