Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylcellulose, 400 cP, USP, 26-33%, Spectrum™ Chemical
SDP

Catalog No. 18604474 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-474 100 g Fiber Drum
18-604-475 500 g Fiber Drum
18-604-476 2.5 kg Fiber Drum
18-604-477 12 kg Fiber Drum
18-604-478 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-604-474 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. ME136100GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

ME136, 9004-67-5

Spectrum™ Chemical Methylcellulose, 400 cP, USP is a bulk-forming laxative. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9004-67-5
Molecular Formula C20H38O11
Molecular Weight (g/mol) 454.51
InChI Key YLGXILFCIXHCMC-UHFFFAOYNA-N
IUPAC Name 3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane
SMILES COCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC

Specifications

CAS Max % 100%
Ignition Residue 1.5%
Loss on Drying 5%
Packaging Fiber Drum
Quantity 100 g
Percent Purity 26 to 33%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Methylcellulose, 400 cP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.