Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methylcobalamin | 13422-55-4 | 99.0% | 1 ML
SDP

Catalog No. 5000408773
Encompass_Preferred

Methylcobalamin, a cobalamin and form of vitamin B12, is characterized by a methyl group replacing the cyanide found in cyanocobalamin. This compound features an octahedral cobalt(III) centre and is known for its rare metal-alkyl bonds. Physiologically, it functions as vitamin B12, addressing deficiencies such as pernicious anemia, and is also applied in treating peripheral neuropathy, diabetic neuropathy, and amyotrophic lateral sclerosis.

  • Form of vitamin B12 (cobalamin)
  • Contains a methyl group instead of cyanide
  • Features an octahedral cobalt(III) centre and metal-alkyl bonds
  • Equivalent to vitamin B12 for treating deficiencies (e.g., pernicious anemia)
  • Used in peripheral neuropathy, diabetic neuropathy, and amyotrophic lateral sclerosis treatment

Catalog No. 50-004-08773 Supplier Medchemexpress LLC Supplier No. HYB058610MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methylcobalamin, a cobalamin and form of vitamin B12, is characterized by a methyl group replacing the cyanide found in cyanocobalamin. This compound features an octahedral cobalt(III) centre and is known for its rare metal-alkyl bonds. Physiologically, it functions as vitamin B12, addressing deficiencies such as pernicious anemia, and is also applied in treating peripheral neuropathy, diabetic neuropathy, and amyotrophic lateral sclerosis.

  • Form of vitamin B12 (cobalamin)
  • Contains a methyl group instead of cyanide
  • Features an octahedral cobalt(III) centre and metal-alkyl bonds
  • Equivalent to vitamin B12 for treating deficiencies (e.g., pernicious anemia)
  • Used in peripheral neuropathy, diabetic neuropathy, and amyotrophic lateral sclerosis treatment

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.