Learn More
Cayman Chemical Methylcobalamin, Purity: ≥98%, CAS: 13422-55-4, 1 g
SDP
Catalog No. 502941911
Encompass_Preferred
An analog of vitamin B12 with diverse neurological activities; promotes neurite outgrowth and survival in primary CGN and DRG cells and activation of ERK1/2 and Akt from 0.1-100 µM; improves sensory function in a pinch test and increases toe spreading in a rat model of sciatic nerve injury at 1 mg/kg per day; decreases the number of atypical mitochondria in the sciatic nerve and reduces vincristine-induced mechanical allodynia and thermal hyperalgesia in a rat model of neuropathic pain; reduces muscle weakness and forelimb contracture and increases bicep muscle weight and the number of musculocutaneous nerves in the wobbler mouse model of ALS; enhances the recovery of compound muscle action potentials and motor end plate innervation and decreases the time to sticker removal in the sticker removal grooming test in a rat model of bicep ulnar to musculocutaneous nerve transfer
Catalog No. 50-294-1911 Supplier Cayman Chemical Supplier No. 291131 G
An analog of vitamin B12 with diverse neurological activities; promotes neurite outgrowth and survival in primary CGN and DRG cells and activation of ERK1/2 and Akt from 0.1-100 µM; improves sensory function in a pinch test and increases toe spreading in a rat model of sciatic nerve injury at 1 mg/kg per day; decreases the number of atypical mitochondria in the sciatic nerve and reduces vincristine-induced mechanical allodynia and thermal hyperalgesia in a rat model of neuropathic pain; reduces muscle weakness and forelimb contracture and increases bicep muscle weight and the number of musculocutaneous nerves in the wobbler mouse model of ALS; enhances the recovery of compound muscle action potentials and motor end plate innervation and decreases the time to sticker removal in the sticker removal grooming test in a rat model of bicep ulnar to musculocutaneous nerve transfer