Learn More
Selleck Chemical LLC Methylcobalamin S4994-25mg
SDP
Catalog No. 503110168
Encompass_Preferred
Methylcobalamin (Mecobalamin Vancomin Methycobal Cobametin) is one active form of vitamin B12 which can directly participate in homocysteine metabolism It is used to treat some nutritional diseases and other diseases in clinic such as Alzheimer s disease and rheumatoid arthritis
Catalog No. 50-311-0168 Supplier Selleck Chemical LLC Supplier No. S499425MG
Methylcobalamin (Mecobalamin Vancomin Methycobal Cobametin) is one active form of vitamin B12 which can directly participate in homocysteine metabolism It is used to treat some nutritional diseases and other diseases in clinic such as Alzheimer s disease and rheumatoid arthritis