Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylcobalamin Dietary Supplement, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18604492 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-492 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-604-492 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M40881GM

missing translation for 'validMainframeMsg'

M4088, 13422-55-4, C63H91CoN13O14P

Spectrum™ Chemical Methylcobalamin, USP Dietary Supplement

Chemical Identifiers

CAS 13422-55-4
Molecular Formula C63H91CoN13O14P
Molecular Weight (g/mol) 1344.41
InChI Key QXOMMGINPOYKPR-UHFFFAOYNA-L
IUPAC Name λ²-cobalt(2+) 8,13,18-tris(2-carbamoylethyl)-3,14,19-tris(carbamoylmethyl)-4-{2-[(2-{[5-(5,6-dimethyl-1H-1,3-benzodiazol-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl phosphonato]oxy}propyl)carbamoyl]ethyl}-1,4,6,9,9,14,16,19-octamethyl-20,21,22,23-tetraazapentacyclo[15.2.1.1²,⁵.1⁷,¹⁰.1¹²,¹⁵]tricosa-5(23),6,10(22),11,15(21),16-hexaen-20-ide methanide
SMILES [CH3-].[Co++].CC(CNC(=O)CCC1(C)C(CC(N)=O)C2N=C1C(C)=C1N=C(C=C3N=C(C(C)=C4[N-]C2(C)C(C)(CC(N)=O)C4CCC(N)=O)C(C)(CC(N)=O)C3CCC(N)=O)C(C)(C)C1CCC(N)=O)OP([O-])(=O)OC1C(CO)OC(C1O)N1C=NC2=CC(C)=C(C)C=C12

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 1 g
Percent Purity 98 to 102%
Grade USP Dietary Supplement
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.