Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylcobalamin Hydrate 98.0+%, TCI America™
SDP

Catalog No. M27421G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M27421G Supplier TCI America Supplier No. M27421G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 288315-09-3
Molecular Formula C63H91CoN13O14P
Molecular Weight (g/mol) 1344.40
MDL Number MFCD00149221
InChI Key XVLRGRFESMDSFT-BVWPOUIRNA-L
Synonym Mecobalamin
PubChem CID 133556284
IUPAC Name (10S,12R,13R,24S,35R,41R)-30,35,40-tris(2-carbamoylethyl)-24,36,41-tris(carbamoylmethyl)-46-hydroxy-12-(hydroxymethyl)-1,5,6,17,23,28,31,31,36,38,41,42-dodecamethyl-15,20-dioxo-11,14,16-trioxa-2λ⁵,9,19,26λ⁵,43,44λ⁵,45λ⁵-heptaaza-15λ⁵-phospha-1-cobaltadodecacyclo[27.14.1.1¹,³⁴.1²,⁹.1¹⁰,¹³.0¹,²⁶.0³,⁸.0²³,²⁷.0²⁵,⁴².0³²,⁴⁴.0³⁹,⁴³.0³⁷,⁴⁵]heptatetraconta-2(47),3,5,7,26,28,32(44),33,37(45),38-decaene-1,2,26,44,45-pentakis(ylium)-15-olate
SMILES C(CC(N)=O)[C@@H]1C(CC(N)=O)(C)C2=[N+]3[Co+]456([N+]7=CN([C@]8(O[C@@H]([C@](OP(OC(CNC(CCC9([C@H](CC(N)=O)C([N+]4=C9C(C)=C4[N+]5=C(C(C4CCC(N)=O)(C)C)C=C13)C1([C@](CC(N)=O)(C(CCC(N)=O)C(=C2C)N61)C)C)C)=O)C)([O-])=O)(C8O)[H])CO)[H])C1=CC(C)=C(C=C71)C)C

Specifications

Color Red
Quantity 1 g
Formula Weight 1344.40
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Methylcobalamin Hydrate
RTECSNumber GG3745000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.