Learn More
Sigma Aldrich Fine Chemicals Biosciences Methylcobalamin vitamin B125MG
Catalog No. 502897599
Encompass
Methylcobalamin (MeCbl) is an analog of vitamin B12 (cobalamin). It differs from cobalamin by containing a methyl group. The physiologically active form of vitamin B12 include methylcobalamin and adenosylcobalamin.
Catalog No. 50-289-7599 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M975625MG
Methylcobalamin (MeCbl) is an analog of vitamin B12 (cobalamin). It differs from cobalamin by containing a methyl group. The physiologically active form of vitamin B12 include methylcobalamin and adenosylcobalamin.