Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MethylCode™ Bisulfite Conversion Kit

Catalog No. 501148733
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-114-8733 50 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-114-8733 Supplier Applied Biosystems™ Supplier No. MECOV50
Add to Cart
Edge
Add to Cart

MethylCode™ Bisulfite Conversion Kit

Please see our newest bisulfite conversion kit, Cells-to-CpG™ Bisulfite Conversion System, designed to provide more flexibility in starting samples without the need to purify your DNA and more confidence with available methylation controls. Cells-to-CpG™ is completely validated for Life Technologies' downstream platforms and replaces MethylCode™ as the bisulfite conversion kit of choice.

The MethylCode™ Bisulfite Conversion Kit is a fast and easy way to convert unmethylated cytosines from a DNA sample into uracils while methylated cytosines remain unchanged.

The modified DNA is ideal for PCR amplification for downstream analyses, including restriction endonuclease digestion, sequencing, and microarrays. With the MethylCode™ Bisulfite Conversion Kit you can expect:

–Simple, rapid cytosine methylation detection

–Consistency across most experimental conditions
–Streamlined conversion of unmethylated cytosines into uracil, thanks to a coupled DNA denaturation/bisulfite treatment step
–Ultra-pure DNA ideal for subsequent molecular-based analyses



Kit specifications:

–Protocol: Rapid heat denaturation of DNA followed by sodium bisulfite modification with spin column purification

–Sample sources: Plasmid, genomic, and endonuclease digested DNAs
–Input DNA: 500 pg-2μg DNA per treatment, with 200-500 ng being optimal
–Recovery volume: Purified, bisulfite-treated DNA is recovered in 10μl elution buffer



Contents and storage:
The MethylCode™ Bisulfite Conversion Kit contains all the reagents necessary to convert your DNA for methylation studies. The kit includes CT Conversion Reagent, Dilution Buffer, Resuspension buffer, Binding Buffer, Wash Buffer, Desulphonation Buffer, Elution Buffer, Spin Columns, Collection Tubes.

Store at room temperature for up to 24 months.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Kit, Bisulfite Conversion
Content And Storage The MethylCode™ Bisulfite Conversion Kit contains all the reagents necessary to convert your DNA for methylation studies. The kit includes CT conversion reagent, dilution buffer, resuspension buffer, binding buffer, wash buffer, desulphonation buffer, elution buffer, spin columns, and collection tubes.
Sample Type DNA (Genomic)
Shipping Condition Room Temperature
Starting Material Cell No. Purified genomic DNA - 200 ng to 500 ng
Format Kit
Product Line MethylCode
Quantity 50 Reactions

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.