Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylcyclohexane, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019272 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9272 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9272 Supplier MilliporeSigma Supelco Supplier No. 662945ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methylcyclohexane [MCH] is a clear organic flammable liquid, mostly used as a solvent. During the development of chemical kinetic mechanisms for other cycloalkanes, MCH is considered as reaction starting point.

Featured Industries: Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 108-87-2
Ignition Point 545°F
Vapor Pressure 83.2 mmHg (37.7°C)
Vapor Density 3.4 (vs. air)
Density 0.77 g/mL (at 25°C)
Flash Point -4.0°C - closed cup
Physical Form Neat
Linear Formula C6H11 CH3
MDL Number MFCD00001497
Melting Point -126°C
Boiling Point 101°C
Synonym Hexahydrotoluene
Type Analytical Standard
Quantity 5 mL
Formula Weight 98.19
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.