Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylcyclopentadiene Dimer (so called) (stabilized with TBC), TCI America™
SDP

Catalog No. M092025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M092025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M092025ML Supplier TCI America Supplier No. M092025ML
Add to Cart
Add to Cart

Chemical Identifiers

Molecular Formula C12H16

Specifications

Color Yellow
Boiling Point 200°C
Quantity 25 mL
UN Number 3295
Physical Form Liquid
Chemical Name or Material Methylcyclopentadiene Dimer (so called) (stabilized with TBC)
EINECSNumber (4)-1808
RTECSNumber PC1075000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.