Learn More
Strem, An Ascensus Company CAS# 12108-13-3. 5g. Methylcyclopentadienylmanganese tricarbonyl, min. 97%. MFCD00001426
SDP
Supplier:  Strem, An Ascensus Company 2515505G
Encompass
CAS# 12108-13-3. 5g. Methylcyclopentadienylmanganese tricarbonyl, min. 97%. MFCD00001426. Molecular Weight: 218.09. Molecular Formula: (CH3C5H4)Mn(CO)3. Color/form: yellow liq. Strem# 25-1550. http://www.strem.com/catalog/v/25-1550/
Catalog No. 50-901-10926