Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylcyclopentane, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019124 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9124 10 mL
11-101-8218 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-9124 Supplier MilliporeSigma Supelco Supplier No. 6649010ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methylcyclopentane, a naphthene hydrocarbon, has a tertiary carbon atom and also has greater reactivity towards chemical agents like sulphuric acid, fuming nitric acid, etc.

Featured Industries: Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 96-37-7
Ignition Point 624°F
Vapor Pressure 232.8 mmHg (37.7°C)
Density 0.748 g/mL (at 20°C)
Flash Point -29.0°C - closed cup
Physical Form Neat
Linear Formula C5H9 CH3
MDL Number MFCD00001382
Melting Point -142°C
Boiling Point 72°C
Type Analytical Standard
Quantity 10 mL
Formula Weight 84.16
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.