Alfa Aesar™ Methyldichlorophosphine, 97+%

Manufacturer: Alfa Aesar™ 3021106

Quantity 5g

 View more versions of this product

Catalog No. AA3021106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 676-83-5
    Linear Formula (CH3)PCl2
    Formula Weight 116.92
    MDL Number MFCD00013616
    UN Number UN2845
    Synonym Methylphosphonous dichloride; Dichloromethylphosphine
    Chemical Name or Material Methyldichlorophosphine

    Specifications

    Formula Weight 116.92
    Health Hazard 3 GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H250-H226-H252-H314-H318
    Catches fire spontaneously if exposed to air.
    Flammable liquid and vapor.
    Self-heating in large quantities
    may catch fire.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Linear Formula (CH3)PCl2
    Odor Strong
    Color Colorless
    Quantity 5g
    Refractive Index 1.4960
    Physical Form Liquid
    UN Number UN2845
    Sensitivity Air & Moisture Sensitive
    Boiling Point 81° to 82°C (729mmHg)
    Synonym Methylphosphonous dichloride; Dichloromethylphosphine
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.
    Density 1.309g/mL
    Flash Point 48° (120°F)
    TSCA TSCA
    MDL Number MFCD00013616
    CAS 676-83-5
    Chemical Name or Material Methyldichlorophosphine
    Assay ≥98%

    GHS H Statement
    H250-H226-H252-H314-H318
    Catches fire spontaneously if exposed to air.
    Flammable liquid and vapor.
    Self-heating in large quantities
    may catch fire.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit