Learn More
Strem, An Ascensus Company CAS# 1486-28-8. 5g. Methyldiphenylphosphine, 99%. MFCD00008508
SDP
Catalog No. 5090110260
Encompass
CAS# 1486-28-8. 5g. Methyldiphenylphosphine, 99%. MFCD00008508. Molecular Weight: 200.22. Molecular Formula: CH3(C6H5)2P. Color/form: colorless liq. Strem# 15-3250. http://www.strem.com/catalog/v/15-3250/
Catalog No. 50-901-10260 Supplier Strem, An Ascensus Company Supplier No. 1532505G
CAS# 1486-28-8. 5g. Methyldiphenylphosphine, 99%. MFCD00008508. Molecular Weight: 200.22. Molecular Formula: CH3(C6H5)2P. Color/form: colorless liq. Strem# 15-3250. http://www.strem.com/catalog/v/15-3250/