Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyldiphenylsilane, 97%

Catalog No. AAL0421106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0421106 5 g
AAL0421114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0421106 Supplier Thermo Scientific Chemicals Supplier No. L0421106
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 776-76-1 | C13H13Si | 197.332 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 776-76-1
Molecular Formula C13H13Si
Molecular Weight (g/mol) 197.332
MDL Number MFCD00008255
InChI Key OKHRRIGNGQFVEE-UHFFFAOYSA-N
Synonym methyldiphenylsilane, diphenylmethylsilane, silane, methyldiphenyl, benzene, 1,1'-methylsilylene bis, methyldiphenylsilyl, methyl diphenyl silicon, methyl-di phenyl silicon, silane, methyldiphenyl-,
PubChem CID 6327660
IUPAC Name methyl(diphenyl)silicon
SMILES C[Si](C1=CC=CC=C1)C2=CC=CC=C2

Specifications

Boiling Point 266°C to 267°C
Quantity 5 g
Beilstein 1867183
Flash Point 120°C (248°F)
Formula Weight 198.34
Percent Purity 97%
Odor Odorless
Refractive Index 1.572
Density 1.001
Chemical Name or Material Methyldiphenylsilane
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 212-281-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.