Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyldiphenylsilane 95.0+%, TCI America™
SDP

Catalog No. D182525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D182525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D182525ML Supplier TCI America Supplier No. D182525ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 776-76-1
Molecular Formula C13H13Si
Molecular Weight (g/mol) 197.332
MDL Number MFCD00008255
InChI Key OKHRRIGNGQFVEE-UHFFFAOYSA-N
Synonym methyldiphenylsilane, diphenylmethylsilane, silane, methyldiphenyl, benzene, 1,1'-methylsilylene bis, methyldiphenylsilyl, methyl diphenyl silicon, methyl-di phenyl silicon, silane, methyldiphenyl-,
PubChem CID 6327660
IUPAC Name methyl(diphenyl)silicon
SMILES C[Si](C1=CC=CC=C1)C2=CC=CC=C2

Specifications

Boiling Point 91°C
Color Yellow
Physical Form Liquid
Quantity 25 mL
Formula Weight 198.34
Percent Purity ≥95.0% (GC)
Chemical Name or Material Methyldiphenylsilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.