Learn More
Apexbio Technology LLC Methyldopa 555-30-6 50mg
SDP
Catalog No. 502240336
Encompass_Preferred
Methyldopa (CAS 555-30-6) is a synthetic small molecule that acts as a centrally acting antihypertensive agent by selectively stimulating 2-adrenergic receptors in the central nervous system This agonist activity inhibits sympathetic outflow resulting in reduced peripheral vascular resistance and lower blood pressure Methyldopa is widely utilized in biomedical research for studying central adrenergic mechanisms sympathetic regulation and the pharmacological modulation of hypertension and related cardiovascular conditions
This item is not returnable. View return policy
Catalog No. 50-224-0336 Supplier Apexbio Technology LLC Supplier No. B178750
Methyldopa (CAS 555-30-6) is a synthetic small molecule that acts as a centrally acting antihypertensive agent by selectively stimulating 2-adrenergic receptors in the central nervous system This agonist activity inhibits sympathetic outflow resulting in reduced peripheral vascular resistance and lower blood pressure Methyldopa is widely utilized in biomedical research for studying central adrenergic mechanisms sympathetic regulation and the pharmacological modulation of hypertension and related cardiovascular conditions