Learn More
Medchemexpress LLC Methyldopate | 6014-30-8 | 98.0% | 1 ML
SDP
Catalog No. 5000412750
Encompass_Preferred
Methyldopate is an ethyl ester proagent of α-Methyldopa (α-MD; HY-B0225) that acts as an α-adrenergic agonist, selective for α2-adrenergic receptors. It has the potential for severe hypertension research and is suitable for laboratory centrifugation and storage. The product is provided as a solid solution.
  • Ethyl ester proagent of α-Methyldopa
  • Acts as an α-adrenergic agonist
  • Selective for α2-adrenergic receptors
  • Potential for severe hypertension research
  • Suitable for laboratory centrifugation and storage
Catalog No. 50-004-12750 Supplier Medchemexpress LLC Supplier No. HYB169610MM/1ML
Methyldopate is an ethyl ester proagent of α-Methyldopa (α-MD; HY-B0225) that acts as an α-adrenergic agonist, selective for α2-adrenergic receptors. It has the potential for severe hypertension research and is suitable for laboratory centrifugation and storage. The product is provided as a solid solution.
  • Ethyl ester proagent of α-Methyldopa
  • Acts as an α-adrenergic agonist
  • Selective for α2-adrenergic receptors
  • Potential for severe hypertension research
  • Suitable for laboratory centrifugation and storage