Learn More
AdipoGen Methyldymron
SDP
Supplier:  AdipoGen CDXM0135M050
Encompass_Preferred
Chemical. CAS 42609-73-4. Formula C17H20N2O. MW 268.4. Synthetic. Herbicide. Mode of action is unknow, but most probably by inhibition of plant cell division. Compound can be used as analytical reference material.
Catalog No. 50-256-0500