Learn More
AdipoGen Methyldymron
SDP
Catalog No. 502560499
Encompass_Preferred
Chemical. CAS 42609-73-4. Formula C17H20N2O. MW 268.4. Synthetic. Herbicide. Mode of action is unknow, but most probably by inhibition of plant cell division. Compound can be used as analytical reference material.
Catalog No. 50-256-0499 Supplier AdipoGen Supplier No. CDXM0135M500
Chemical. CAS 42609-73-4. Formula C17H20N2O. MW 268.4. Synthetic. Herbicide. Mode of action is unknow, but most probably by inhibition of plant cell division. Compound can be used as analytical reference material.