Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyle1-methyl-1H-imidazole-5-carboxylate, 97%, Thermo Scientific™

Catalog No. TL00659DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
TL00659DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TL00659DA Supplier Thermo Scientific Chemicals Supplier No. TL00659DA
Add to Cart

Chemical Identifiers

CAS 17289-20-2
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.14
MDL Number MFCD01567300
InChI Key AKDPLDCXQNEMCL-UHFFFAOYSA-N
Synonym methyl 1-methyl-1h-imidazole-5-carboxylate, methyl 1-methylimidazole-5-carboxylate, methyl1-methyl-1h-imidazole-5-carboxylate, 3-methyl-3h-imidazole-4-carboxylic acid methyl ester, 1h-imidazole-5-carboxylic acid, 1-methyl-, methyl ester, 1-methyl-1h-imidazole-5-carboxylic acid methyl ester, pubchem9004, acmc-209e5h, 1-methylimidazole-5-carboxylic acid methyl ester, 1h-imidazole-5-carboxylicacid, 1-methyl-, methyl ester
PubChem CID 2736896
IUPAC Name methyl 3-methylimidazole-4-carboxylate
SMILES COC(=O)C1=CN=CN1C

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 140.14
Percent Purity 97%
Chemical Name or Material Methyle1-methyl-1H-imidazole-5-carboxylate
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.