Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyle2-(2,5-dimethyl-1,3-thiazol-4-yl)acetate, 97%, Thermo Scientific™

Catalog No. TL01020DA
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
TL01020DA 1 g Amber Glass Bottle
TL01020EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. TL01020DA Supplier Thermo Scientific Chemicals Supplier No. TL01020DA
Add to Cart

Chemical Identifiers

CAS 306937-37-1
Molecular Formula C8H11NO2S
Molecular Weight (g/mol) 185.241
MDL Number MFCD02677686
InChI Key AUEJOXGQRQYLDJ-UHFFFAOYSA-N
Synonym methyl 2-2,5-dimethyl-1,3-thiazol-4-yl acetate, methyl 2-2,5-dimethylthiazol-4-yl acetate, methyl 2,5-dimethyl-1,3-thiazol-4-yl acetate, methyl 2-dimethyl-1,3-thiazol-4-yl acetate, acmc-1agna, 4-thiazoleaceticacid, 2,5-dimethyl-, methyl ester, 2,5-dimethyl-thiazol-4-yl-acetic acid methyl ester, 2-2,5-dimethyl-4-thiazolyl acetic acid methyl ester, methyl 2-2,5-dimethyl-1,3-thiazol-4-yl ethanoate
PubChem CID 2801571
IUPAC Name methyl 2-(2,5-dimethyl-1,3-thiazol-4-yl)acetate
SMILES CC1=C(N=C(S1)C)CC(=O)OC

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 185.25
Percent Purity 97%
Chemical Name or Material Methyle2-(2, 5-dimethyl-1, 3-thiazol-4-yl)acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.