Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyle3-amino-5-(tert-butyl)thiophene-2-carboxylate, 97%, Thermo Scientific™

Catalog No. GK00671DE
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
GK00671DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. GK00671DE Supplier Thermo Scientific Chemicals Supplier No. GK00671DE
Add to Cart

Chemical Identifiers

CAS 175137-03-8
Molecular Formula C10H15NO2S
Molecular Weight (g/mol) 213.295
MDL Number MFCD00068163
InChI Key WHJBGOQJMFAKHY-UHFFFAOYSA-N
Synonym methyl 3-amino-5-tert-butyl thiophene-2-carboxylate, 3-amino-5-tert-butyl-thiophene-2-carboxylic acid methyl ester, methyl 3-amino-5-4-tert-butyl thiophene-2-carboxylate, maybridge1_003935, methyl 3-amino-5-tert-butyl-2-thiophenecarboxylate, 3-amino-5-tert-butyl-2-thiophenecarboxylic acid methyl ester, 3-amino-5-tert-butylthiophene-2-carboxylic acid methyl ester, 2-thiophenecarboxylicacid, 3-amino-5-1,1-dimethylethyl-, methyl ester
PubChem CID 2777503
IUPAC Name methyl 3-amino-5-tert-butylthiophene-2-carboxylate
SMILES CC(C)(C)C1=CC(=C(S1)C(=O)OC)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 213.3
Percent Purity 97%
Chemical Name or Material Methyl 3-amino-5-(tert-butyl)thiophene-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.