Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyle3-hydroxybenzo[b]thiophene-2-carboxylate, 97%, Thermo Scientific™

Catalog No. BTB04875EA
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB04875EA 10 g Amber Glass Bottle
BTB04875DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTB04875EA Supplier Thermo Scientific Chemicals Supplier No. BTB04875EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13134-76-4
Molecular Formula C10H8O3S
Molecular Weight (g/mol) 208.23
MDL Number MFCD00067732
InChI Key JVUOTPTULVZUTL-KTKRTIGZSA-N
Synonym methyl 3-hydroxybenzo b thiophene-2-carboxylate, methyl 3-hydroxybenzo b thiophene-3-carboxylate, 2-hydroxy methoxy methylidene-1-benzothiophen-3-one, 2z-2-hydroxy methoxy methylidene-1-benzothiophen-3-one, benzo b thiophene-2-carboxylicacid, 3-hydroxy-, methyl ester, benzo b thiophene-2-carboxylic acid, 3-hydroxy-, methyl ester, maybridge4_000295, methyl 3-hydroxy-benzothiophene2-carboxylate
PubChem CID 83165
SMILES CO\C(O)=C1/SC2=CC=CC=C2C1=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
IUPAC Name (2Z)-2-[hydroxy(methoxy)methylidene]-2,3-dihydro-1-benzothiophen-3-one
Formula Weight 208.24
Percent Purity 97%
Chemical Name or Material Methyl 3-hydroxybenzo[b]thiophene-2-carboxylate
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.