Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

methyle4-aminothiophene-3-carboxylate hydrochloride, 97%, Thermo Scientific™

Catalog No. GK01904EB
Change view
Click to view available options
Quantity:
1 g
25 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
GK01904EB 25 g Amber Glass Bottle
GK01904DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. GK01904EB Supplier Thermo Scientific Chemicals Supplier No. GK01904EB
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 39978-14-8
Molecular Formula C6H7NO2S
Molecular Weight (g/mol) 157.19
MDL Number MFCD00068149
InChI Key BUFZZXCVOFBHLS-UHFFFAOYSA-N
Synonym methyl 4-aminothiophene-3-carboxylate hydrochloride, methyl 3-aminothiophene-4-carboxylate hydrochloride, 3-carbomethoxy-4-aminothiophene hydrochloride, acmc-209zkz, methyl 4-aminothiophene-3-carboxylate, chloride, 3-amino-4-methoxycarbonylthiophene hydrochloride, 4-amino-3-methoxycarbonylthiophene hydrochloride, methyl3-aminothiophene-4-carboxylatehydrochloride, 4-amino-3-methoxycarbonyl thiophene hydrochloride, methyl 4-aminothiophen-3-carboxylate hydrochloride
PubChem CID 2777609
IUPAC Name methyl 4-aminothiophene-3-carboxylate;hydrochloride
SMILES COC(=O)C1=CSC=C1N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 25 g
Formula Weight 193.65
Percent Purity 97%
Chemical Name or Material Methyl 4-aminothiophene-3-carboxylate hydrochloride
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.