Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyle6-piperidinonicotinate, 97%, Thermo Scientific™

Catalog No. MO07008EA
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO07008EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO07008EA Supplier Thermo Scientific Chemicals Supplier No. MO07008EA

Chemical Identifiers

CAS 132521-82-5
Molecular Formula C12H16N2O2
Molecular Weight (g/mol) 220.272
InChI Key UJSPHYCMAZIKQD-UHFFFAOYSA-N
Synonym methyl 6-piperidinonicotinate, methyl 6-piperidin-1-yl nicotinate, methyl 6-piperidin-1-yl pyridine-3-carboxylate, methyl 6-1-piperidyl nicotinate, methyl 6-piperidylpyridine-3-carboxylate, methyl 6-piperidin-1-ylnicotinate
PubChem CID 2794747
IUPAC Name methyl 6-piperidin-1-ylpyridine-3-carboxylate
SMILES COC(=O)C1=CN=C(C=C1)N2CCCCC2

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 220.27
Chemical Name or Material Methyle6-piperidinonicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.