Learn More
Crescent Chemical Co Inc METHYLENE BISACRYLAMIDE 2X
SDP
Catalog No. 50247858
Encompass_Preferred
Methylene bisacrylamide 2X solution 2 % (w/v) N,N'-Methylene bisacrylamide 2X solution. Solution contains 2 % (w/v) highly purified »Bis« in deionized water. Size - 1L Storage Conditions - +2 °C TO +8 °C Catalog Number - 29197.01
Catalog No. 50-247-858 Supplier Crescent Chemical Co Inc Supplier No. SE29197.01
Methylene bisacrylamide 2X solution 2 % (w/v) N,N'-Methylene bisacrylamide 2X solution. Solution contains 2 % (w/v) highly purified »Bis« in deionized water. Size - 1L Storage Conditions - +2 °C TO +8 °C Catalog Number - 29197.01