Learn More
Methylene Blue, 0.5% w/v, Aqueous ASTM C837, Reagents
Specifications
Specifications
| Chemical Name or Material | Methylene Blue, 0.5% w/v, Aqueous ASTM C837 |
| CAS | 7732-18-5, 7220-79-3 |
| Boiling Point | 100°C |
| Packaging | Glass Bottle |
| Specific Gravity | 1.0002 |
| Synonym | 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride Trihydrate, Basic Blue 9 Trihydrate, Color Index 52015 |
| Color | Blue |
| Quantity | 250 mL |
| Physical Form | Liquid |
| Percent Purity | 0.5% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.