Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Blue, 1% (w/v) Aqueous Solution, Spectrum™ Chemical
SDP

Catalog No. 18611764 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-611-764 125 mL
18-611-766 500 mL
18-611-765 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-611-764 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M204125ML
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Methylene Blue
CAS 61-73-4, 7732-18-5, 7220-79-3
Concentration 1%
Grade Chemical Solution
Packaging Poly Bottle
Color Blue
Quantity 125 mL

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.