Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Blue, 1% (w/v) in Denatured Alcohol, Ricca Chemical
SDP

Catalog No. 48644 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
4864-4 120 mL Natural Poly Bottle
4864-16 500 mL Natural Poly Bottle
486432 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 4864-4 Supplier Ricca Chemical Company Supplier No. 48644
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Methylene Blue
CAS 64-17-5, 67-56-1, 7220-79-3
Packaging Natural Poly Bottle
Molecular Formula CH4O
MDL Number MFCD00004595
Color Blue
Quantity 120 mL
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) Mixture
Percent Purity 1% W/V
Grade Laboratory
Concentration or Composition (by Analyte or Components) 1% (w/v) Alcoholic Solution, in SDA-3A (100 parts Ethanol and 5 parts Methanol)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.