Learn More
Medchemexpress LLC HY-14536 100mg Medchemexpress, Methylene Blue CAS:61-73-4 Purity:>98%
SDP
Catalog No. 502030541
Encompass_Preferred
Medchemexpress, HY-14536 100mg Methylene Blue CAS:61-73-4 Methylene blue (Basic Blue 9) is a guanylyl cyclase (sGC), monoamine oxidase A (MAO-A) and NO synthase (NOS) inhibitor. Methylene blue is a vasopressor and is often used as a dye in several medical procedures. Methylene blue has antinociception, antimalarial, antidepressant and anxiolytic activity effects. Methylene Blue has the potential for methemoglobinemias, neurodegenerative disorders and ifosfamide-induced encephalopathytreatment[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0541 Supplier Medchemexpress LLC Supplier No. HY14536100MG
Medchemexpress, HY-14536 100mg Methylene Blue CAS:61-73-4 Methylene blue (Basic Blue 9) is a guanylyl cyclase (sGC), monoamine oxidase A (MAO-A) and NO synthase (NOS) inhibitor. Methylene blue is a vasopressor and is often used as a dye in several medical procedures. Methylene blue has antinociception, antimalarial, antidepressant and anxiolytic activity effects. Methylene Blue has the potential for methemoglobinemias, neurodegenerative disorders and ifosfamide-induced encephalopathytreatment[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.