Learn More
Crescent Chemical Co Inc METHYLENE BLUE
SDP
Catalog No. 50247859
Encompass_Preferred
Methylene Blue Standardized according to DIN 58981. Methylene blue is very sensitive to oxidation and usually contains demethylated products, called Azure A, B and C and thionine. For staining and detecting RNA in PAGE (as 0.1 % solution in water). Water solubl
Catalog No. 50-247-859 Supplier Crescent Chemical Co Inc Supplier No. SE29198.01
Methylene Blue Standardized according to DIN 58981. Methylene blue is very sensitive to oxidation and usually contains demethylated products, called Azure A, B and C and thionine. For staining and detecting RNA in PAGE (as 0.1 % solution in water). Water solubl